Anti-UBOX5 Mouse Monoclonal Antibody [clone: 1C5]

Supplier: OriGene
ORIGCF503340EA 749 EUR
ORIGCF503340
Anti-UBOX5 Mouse Monoclonal Antibody [clone: 1C5]
Antibodies
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: UBOX5
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR