Anti-IRF6 Mouse Monoclonal Antibody [clone: 2A12]

Supplier: OriGene
ORIGCF503345EA 749 EUR
ORIGCF503345
Anti-IRF6 Mouse Monoclonal Antibody [clone: 2A12]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: IRF6
Clonality: Monoclonal
Clone: 2A12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR