Anti-ERI1 Mouse Monoclonal Antibody [clone: 1B3]

Supplier: OriGene
ORIGCF504134EA 749 EUR
ORIGCF504134
Anti-ERI1 Mouse Monoclonal Antibody [clone: 1B3]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: ERI1
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR