Anti-AGPAT5 Mouse Monoclonal Antibody [clone: 4H3]

Supplier: OriGene
ORIGCF504145EA 749 EUR
ORIGCF504145
Anti-AGPAT5 Mouse Monoclonal Antibody [clone: 4H3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: AGPAT5
Clonality: Monoclonal
Clone: 4H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR