Anti-TULP3 Mouse Monoclonal Antibody [clone: 6D8]

Supplier: OriGene
ORIGCF504154EA 770 EUR
ORIGCF504154
Anti-TULP3 Mouse Monoclonal Antibody [clone: 6D8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: TULP3
Clonality: Monoclonal
Clone: 6D8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR