Anti-SOCS3 Mouse Monoclonal Antibody [clone: 6F3]

Supplier: OriGene
ORIGCF502922EA 749 EUR
ORIGCF502922
Anti-SOCS3 Mouse Monoclonal Antibody [clone: 6F3]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: SOCS3
Clonality: Monoclonal
Clone: 6F3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR