Anti-RAB30 Mouse Monoclonal Antibody [clone: 4B10]

Supplier: OriGene
ORIGCF505342EA 749 EUR
ORIGCF505342
Anti-RAB30 Mouse Monoclonal Antibody [clone: 4B10]
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RAB30
Clonality: Monoclonal
Clone: 4B10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR