Anti-NMT2 Mouse Monoclonal Antibody [clone: 1G2]

Supplier: OriGene
ORIGCF504199EA 770 EUR
ORIGCF504199
Anti-NMT2 Mouse Monoclonal Antibody [clone: 1G2]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NMT2
Clonality: Monoclonal
Clone: 1G2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR