Anti-STAT4 Mouse Monoclonal Antibody [clone: 2B4]

Supplier: OriGene
ORIGCF502982EA 770 EUR
ORIGCF502982
Anti-STAT4 Mouse Monoclonal Antibody [clone: 2B4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: STAT4
Clonality: Monoclonal
Clone: 2B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR