Anti-LGALS3BP Mouse Monoclonal Antibody [clone: 1F8]

Supplier: OriGene
ORIGCF503454EA 749 EUR
ORIGCF503454
Anti-LGALS3BP Mouse Monoclonal Antibody [clone: 1F8]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LGALS3BP
Clonality: Monoclonal
Clone: 1F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR