Anti-ALG2 Mouse Monoclonal Antibody [clone: 1E7]

Supplier: OriGene
ORIGCF504218EA 749 EUR
ORIGCF504218
Anti-ALG2 Mouse Monoclonal Antibody [clone: 1E7]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: ALG2
Clonality: Monoclonal
Clone: 1E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR