Anti-HDHD2 Mouse Monoclonal Antibody [clone: 2E9]

Supplier: OriGene
ORIGCF503014EA 749 EUR
ORIGCF503014
Anti-HDHD2 Mouse Monoclonal Antibody [clone: 2E9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: HDHD2
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR