Anti-KIF25 Mouse Monoclonal Antibody [clone: 2G9]

Supplier: OriGene
ORIGCF505429EA 770 EUR
ORIGCF505429
Anti-KIF25 Mouse Monoclonal Antibody [clone: 2G9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: KIF25
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR