Anti-COG6 Mouse Monoclonal Antibody [clone: 4G4]

Supplier: OriGene
ORIGCF505449EA 749 EUR
ORIGCF505449
Anti-COG6 Mouse Monoclonal Antibody [clone: 4G4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: COG6
Clonality: Monoclonal
Clone: 4G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR