Anti-RFXANK Mouse Monoclonal Antibody [clone: 2B6]

Supplier: OriGene
ORIGCF504274EA 749 EUR
ORIGCF504274
Anti-RFXANK Mouse Monoclonal Antibody [clone: 2B6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: RFXANK
Clonality: Monoclonal
Clone: 2B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR