Anti-FMR1 Mouse Monoclonal Antibody [clone: 2C3]

Supplier: OriGene
ORIGCF504293EA 749 EUR
ORIGCF504293
Anti-FMR1 Mouse Monoclonal Antibody [clone: 2C3]
Antibodies
Recommended Dilutions: Western Blot: 1:200-1000; Immunochemistry: 1:150

Type: Primary
Antigen: FMR1
Clonality: Monoclonal
Clone: 2C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR