Anti-CDK2 Mouse Monoclonal Antibody [clone: 5A10]

Supplier: OriGene
ORIGCF503104EA 749 EUR
ORIGCF503104
Anti-CDK2 Mouse Monoclonal Antibody [clone: 5A10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CDK2
Clonality: Monoclonal
Clone: 5A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR