Anti-GADD45G Mouse Monoclonal Antibody [clone: 1D10]

Supplier: OriGene
ORIGCF505484EA 749 EUR
ORIGCF505484
Anti-GADD45G Mouse Monoclonal Antibody [clone: 1D10]
Antibodies
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: GADD45G
Clonality: Monoclonal
Clone: 1D10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR