Anti-ERBB2 Mouse Monoclonal Antibody [clone: 3F11]

Supplier: OriGene
ORIGCF503525EA 749 EUR
ORIGCF503525
Anti-ERBB2 Mouse Monoclonal Antibody [clone: 3F11]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: 3F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR