Anti-PPARA Mouse Monoclonal Antibody [clone: 2G6]

Supplier: OriGene
ORIGCF503545EA 749 EUR
ORIGCF503545
Anti-PPARA Mouse Monoclonal Antibody [clone: 2G6]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PPARA
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR