Anti-MTFMT Mouse Monoclonal Antibody [clone: 1E7]

Supplier: OriGene
ORIGCF503565EA 749 EUR
ORIGCF503565
Anti-MTFMT Mouse Monoclonal Antibody [clone: 1E7]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: MTFMT
Clonality: Monoclonal
Clone: 1E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR