Anti-DTNB Mouse Monoclonal Antibody [clone: 2E4]

Supplier: OriGene
ORIGCF504320EA 749 EUR
ORIGCF504320
Anti-DTNB Mouse Monoclonal Antibody [clone: 2E4]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DTNB
Clonality: Monoclonal
Clone: 2E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR