Anti-LCMT1 Mouse Monoclonal Antibody [clone: 2C9]

Supplier: OriGene
ORIGCF503125EA 749 EUR
ORIGCF503125
Anti-LCMT1 Mouse Monoclonal Antibody [clone: 2C9]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: LCMT1
Clonality: Monoclonal
Clone: 2C9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR