Anti-ILVBL Mouse Monoclonal Antibody [clone: 6G9]

Supplier: OriGene
ORIGCF503128EA 749 EUR
ORIGCF503128
Anti-ILVBL Mouse Monoclonal Antibody [clone: 6G9]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: ILVBL
Clonality: Monoclonal
Clone: 6G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR