Anti-ILVBL Mouse Monoclonal Antibody [clone: 3E9]

Supplier: OriGene
ORIGCF503141EA 749 EUR
ORIGCF503141
Anti-ILVBL Mouse Monoclonal Antibody [clone: 3E9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: ILVBL
Clonality: Monoclonal
Clone: 3E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR