Anti-TAPBPL Mouse Monoclonal Antibody [clone: 1C9]

Supplier: OriGene
ORIGCF505532EA 749 EUR
ORIGCF505532
Anti-TAPBPL Mouse Monoclonal Antibody [clone: 1C9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:100

Type: Primary
Antigen: TAPBPL
Clonality: Monoclonal
Clone: 1C9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR