Anti-PRMT2 Mouse Monoclonal Antibody [clone: 3B3]

Supplier: OriGene
ORIGCF503588EA 770 EUR
ORIGCF503588
Anti-PRMT2 Mouse Monoclonal Antibody [clone: 3B3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PRMT2
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR