Anti-BCAT1 Mouse Monoclonal Antibody [clone: 3F5]

Supplier: OriGene
ORIGCF504360EA 749 EUR
ORIGCF504360
Anti-BCAT1 Mouse Monoclonal Antibody [clone: 3F5]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: BCAT1
Clonality: Monoclonal
Clone: 3F5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR