Anti-ERCC1 Mouse Monoclonal Antibody [clone: 1C10]

Supplier: OriGene
ORIGCF504365EA 749 EUR
ORIGCF504365
Anti-ERCC1 Mouse Monoclonal Antibody [clone: 1C10]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC1
Clonality: Monoclonal
Clone: 1C10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR