Anti-IRF6 Mouse Monoclonal Antibody [clone: 2C10]

Supplier: OriGene
ORIGCF503159EA 749 EUR
ORIGCF503159
Anti-IRF6 Mouse Monoclonal Antibody [clone: 2C10]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: IRF6
Clonality: Monoclonal
Clone: 2C10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR