Anti-TMOD1 Mouse Monoclonal Antibody [clone: 2C2]

Supplier: OriGene
ORIGCF503170EA 749 EUR
ORIGCF503170
Anti-TMOD1 Mouse Monoclonal Antibody [clone: 2C2]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: TMOD1
Clonality: Monoclonal
Clone: 2C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR