Anti-TMEFF2 Mouse Monoclonal Antibody [clone: 1A9]

Supplier: OriGene
ORIGCF504426EA 749 EUR
ORIGCF504426
Anti-TMEFF2 Mouse Monoclonal Antibody [clone: 1A9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TMEFF2
Clonality: Monoclonal
Clone: 1A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR