Anti-SHPK Mouse Monoclonal Antibody [clone: 1D2]

Supplier: OriGene
ORIGCF503209EA 749 EUR
ORIGCF503209
Anti-SHPK Mouse Monoclonal Antibody [clone: 1D2]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: SHPK
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR