Anti-BID Mouse Monoclonal Antibody [clone: 3E9]

Supplier: OriGene
ORIGCF506766EA 770 EUR
ORIGCF506766
Anti-BID Mouse Monoclonal Antibody [clone: 3E9]
Antibodies
Recommended Dilutions: Western Blot: 1:400-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: BID
Clonality: Monoclonal
Clone: 3E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR