Anti-TUBB1 Mouse Monoclonal Antibody [clone: 4A3]

Supplier: OriGene
ORIGCF506805EA 749 EUR
ORIGCF506805
Anti-TUBB1 Mouse Monoclonal Antibody [clone: 4A3]
Antibodies
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: TUBB1
Clonality: Monoclonal
Clone: 4A3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR