Anti-EZH2 Mouse Monoclonal Antibody [clone: 2B10]

Supplier: OriGene
ORIGCF802907EA 749 EUR
ORIGCF802907
Anti-EZH2 Mouse Monoclonal Antibody [clone: 2B10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: EZH2
Clonality: Monoclonal
Clone: 2B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR