Anti-NAB1 Mouse Monoclonal Antibody [clone: 5B11]

Supplier: OriGene
ORIGCF802910EA 749 EUR
ORIGCF802910
Anti-NAB1 Mouse Monoclonal Antibody [clone: 5B11]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NAB1
Clonality: Monoclonal
Clone: 5B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR