Anti-CD68 Mouse Monoclonal Antibody [clone: 10H10]

Supplier: OriGene
ORIGCF802964EA 749 EUR
ORIGCF802964
Anti-CD68 Mouse Monoclonal Antibody [clone: 10H10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: 10H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR