Anti-ALB Mouse Monoclonal Antibody [clone: 4D11]
Supplier: OriGene
ORIGCF507287EA 749 EUR
ORIGCF507287
Anti-ALB Mouse Monoclonal Antibody [clone: 4D11]
Antibodies
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: ALB
Clonality: Monoclonal
Clone: 4D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human