Imprimer

Anti-SYTL5 Rabbit Polyclonal Antibody

Fournisseur: Abnova
ABNOPAB21790EA 518 EUR
ABNOPAB21790
Anti-SYTL5 Rabbit Polyclonal Antibody
Anticorps
Rabbit polyclonal antibody raised against recombinant SYTL5.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500

Type: Primary
Antigen: SYTL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR