Anti-TTFC Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11379
ABNOPAB11379EA 772 EUR
ABNOPAB11379
Anti-TTFC Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant TTFC.

Recommended Dilutions: ELISA: 1:100000-1:10000000; Western Blot: 1:20000-1:100000

Type: Primary
Antigen: TTFC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Clostridium Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR