Anti-ALK Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB25504EA 399 EUR
ABNOPAB25504
Anti-ALK Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ALK.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: ALK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR