Anti-Flag tag Polyclonal Antibody

Supplier: Abnova
PAB25505
ABNOPAB25505EA 399 EUR
ABNOPAB25505
Anti-Flag tag Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Flag tag.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: Flag tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR