Anti-ADORA1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB25541
ABNOPAB25541EA 643 EUR
ABNOPAB25541
Anti-ADORA1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ADORA1.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 7.2 ?g/ml

Type: Primary
Antigen: ADORA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR