Anti-CASP3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB25662
ABNOPAB25662EA 518 EUR
ABNOPAB25662
Anti-CASP3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CASP3.

Recommended Dilutions: Immunohistochemistry: 5 ?g/ml

Type: Primary
Antigen: CASP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR