Imprimer

Anti-DRGX Chicken Polyclonal Antibody

Fournisseur: Abnova
PAB25731
ABNOPAB25731EA 518 EUR
ABNOPAB25731
Anti-DRGX Chicken Polyclonal Antibody
Anticorps
Chicken polyclonal antibody raised against synthetic peptide of DRGX.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: DRGX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR