Anti-SPATA19 Chicken Polyclonal Antibody

Supplier: Abnova
PAB25772
ABNOPAB25772EA 518 EUR
ABNOPAB25772
Anti-SPATA19 Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal antibody raised against synthetic peptide of SPATA18.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: SPATA19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR