Anti-PTCHD2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB25778
ABNOPAB25778EA 518 EUR
ABNOPAB25778
Anti-PTCHD2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of PTCHD2.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry: 2.5 ?g/ml

Type: Primary
Antigen: PTCHD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR