Anti-CDC20 Mouse Monoclonal Antibody [clone: AR12]

Supplier: Abnova
MAB11369
ABNOMAB11369EA 518 EUR
ABNOMAB11369
Anti-CDC20 Mouse Monoclonal Antibody [clone: AR12]
Antibodies
Mouse monoclonal antibody raised against CDC20.

Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/million cells; Immunofluorescence: 0.5-1 ?g/ml; Immunohistochemistry: 0.5-1 ?g/ml for 30 min at RT; Immunoprecipitation: 0.5-1 ?g/500 ?g protein lysate; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CDC20
Clonality: Monoclonal
Clone: AR12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR