Anti-AMACR Rabbit Monoclonal Antibody [clone: 13H4]

Supplier: Abnova
MAB11353
ABNOMAB11353EA 518 EUR
ABNOMAB11353
Anti-AMACR Rabbit Monoclonal Antibody [clone: 13H4]
Antibodies
Mouse monoclonal antibody raised against full length recombinant AMACR.

Recommended Dilutions: Immunohistochemistry: 1:100-1:200 for 30 min at RT; Western Blot: 1:100-1:200 for 2 hours at RT)

Type: Primary
Antigen: AMACR
Clonality: Monoclonal
Clone: 13H4
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR